Assay Details
| Gene Symbol: Mirlet7c-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031804.1 | 1 | 48 | 81 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | FQ145175.1 | - | Chr. - - [-] on Build | 81 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | proline rich 34 | Prr34 | -50.2kb | Anti-sense | Chr. 7 - 126536246 - 126540039 on NCBI Build Rnor_6.0 | |
| Downstream | microRNA let-7b | Mirlet7b | rno-let-7b | 321.0 | Sense | Chr. 7 - 126590627 - 126590711 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mirlet7c-2
Genomic Map