Assay Details
| Gene Symbol: Mir221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031934.1 | 1 | 48 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 222 | Mir222 | rno-mir-222 | -459.0 | Sense | Chr. X - 3683919 - 3684021 on NCBI Build Rnor_6.0 |
| Downstream | similar to RIKEN cDNA 4930578C19 | RGD1559731 | 1.0mb | Sense | Chr. X - 4691406 - 4743999 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir221
Genomic Map