Assay Details
| Gene Symbol: Mir135a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031881.1 | 1 | 47 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thymopoietin | Tmpo | LAP2 | -1.0mb | Sense | Chr. 7 - 31847412 - 31872416 on NCBI Build Rnor_6.0 |
| Downstream |
neural precursor cell expressed, developmentally down-regulated 1 |
Nedd1 | 649.1kb | Sense | Chr. 7 - 33544097 - 33584406 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir135a
Genomic Map