Assay Details
| Gene Symbol: Mir490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032737.1 | 1 | 40 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myotrophin | Mtpn | Gcdp | -303.5kb | Anti-sense | Chr. 4 - 63012009 - 63039422 on NCBI Build Rnor_6.0 |
| Downstream |
cholinergic receptor, muscarinic 2 |
Chrm2 | Acm2 | 746.0kb | Sense | Chr. 4 - 64089028 - 64091090 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir490
Genomic Map