Assay Details
| Gene Symbol: Mir138-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031884.1 | 1 | 46 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 12 (sodium/chloride transporter), member 3 |
Slc12a3 |
NCC, TSC |
-5.1kb | Sense | Chr. 19 - 11106033 - 11144674 on NCBI Build Rnor_6.0 |
| Downstream | nucleoporin 93 | Nup93 | 9.9kb | Sense | Chr. 19 - 11159769 - 11263980 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir138-2
Genomic Map