Assay Details
| Gene Symbol: Mir185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031903.1 | 1 | 37 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transport and golgi organization 2 homolog (Drosophila) | Tango2 | RGD1310348 | Sense | Chr. 11 - 86793961 - 86840556 on NCBI Build Rnor_6.0 | |
| Downstream | DiGeorge syndrome critical region gene 8 | Dgcr8 | 39.9kb | Sense | Chr. 11 - 86852676 - 86885233 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir185
Genomic Map