Assay Details
| Gene Symbol: Mir205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031920.1 | 1 | 56 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mucin-5AC-like | LOC102551336 | -1.7kb | Anti-sense | Chr. 13 - 112226490 - 112231626 on NCBI Build Rnor_6.0 | |
| Downstream | plexin A2 | Plxna2 | 1.1mb | Anti-sense | Chr. 13 - 113373569 - 113570774 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir205
Genomic Map