Assay Details
| Gene Symbol: Mir222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031935.1 | 1 | 46 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 7 | Chst7 | Gn6st-4 | -1.0mb | Anti-sense | Chr. X - 2623018 - 2657155 on NCBI Build Rnor_6.0 |
| Downstream | microRNA 221 | Mir221 | rno-mir-221 | 459.0 | Sense | Chr. X - 3684480 - 3684588 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir222
Genomic Map