Assay Details
| Gene Symbol: Mir540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032118.1 | 1 | 43 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-3544 | Mir3544 | rno-mir-3544 | -183.0 | Anti-sense | Chr. 6 - 133705932 - 133706019 on NCBI Build Rnor_6.0 |
| Downstream | microRNA 665 | Mir665 | rno-mir-665 | 172.0 | Sense | Chr. 6 - 133706457 - 133706550 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir540
Genomic Map