Assay Details
| Gene Symbol: Mir336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031780.1 | 1 | 47 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glutamine-fructose-6-phosphate transaminase 2 | Gfpt2 | -24.3kb | Sense | Chr. 10 - 35279237 - 35325506 on NCBI Build Rnor_6.0 | |
| Downstream |
RasGEF domain family, member 1C |
Rasgef1c | 42.9kb | Sense | Chr. 10 - 35392762 - 35459267 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir336
Genomic Map