Assay Details
| Gene Symbol: Mir349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031791.1 | 1 | 43 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase 11 | Mapk11 | p38beta | -17.9kb | Sense | Chr. 7 - 130121678 - 130128633 on NCBI Build Rnor_6.0 |
| Downstream | DENN/MADD domain containing 6B | Dennd6b |
Fam116b, RGD1308012 |
16.1kb | Sense | Chr. 7 - 130162728 - 130178277 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir349
Genomic Map