Assay Details
| Gene Symbol: Mir7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031810.1 | 1 | 47 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fem-1 homolog a (C. elegans) | Fem1a | -11.3kb | Sense | Chr. 9 - 10798934 - 10801140 on NCBI Build Rnor_6.0 | |
| Downstream | dipeptidyl peptidase 9 | Dpp9 | 34.0kb | Anti-sense | Chr. 9 - 10846493 - 10883004 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7b
Genomic Map