Assay Details
| Gene Symbol: Mir871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032255.1 | 1 | 32 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-881 | Mir881 | rno-mir-881 | -3.8kb | Sense | Chr. X - 151283910 - 151283986 on NCBI Build Rnor_6.0 |
| Downstream | microRNA mir-3580 | Mir3580 | rno-mir-3580 | 7.8kb | Sense | Chr. X - 151295615 - 151295695 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir871
Genomic Map