Assay Details
| Gene Symbol: Mir874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032257.1 | 1 | 39 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hypothetical protein LOC681410 | LOC681410 | -112.9kb | Sense | Chr. 17 - 6909501 - 6912145 on NCBI Build Rnor_6.0 | |
| Downstream | uncharacterized LOC102549404 | LOC102549404 | 479.5kb | Anti-sense | Chr. 17 - 7504595 - 7510945 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir874
Genomic Map