Assay Details
| Gene Symbol: Mir196c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032269.1 | 1 | 39 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100911410 | LOC100911410 | Sense | Chr. 10 - 84099076 - 84117851 on NCBI Build Rnor_6.0 | ||
| Downstream | homeo box B9 | Hoxb9 | 6.2kb | Sense | Chr. 10 - 84119884 - 84123390 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir196c
Genomic Map