Assay Details
| Gene Symbol: Mir455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032279.1 | 1 | 37 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 12 | Kif12 | -88.8kb | Anti-sense | Chr. 5 - 79001708 - 79008365 on NCBI Build Rnor_6.0 | |
| Downstream | orosomucoid 1 | Orm1 |
AGP, Agpa1, OMD, Orm |
82.4kb | Sense | Chr. 5 - 79179668 - 79182820 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir455
Genomic Map