Assay Details
| Gene Symbol: Mir652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032300.1 | 1 | 34 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | acyl-CoA synthetase long-chain family member 4 | Acsl4 |
Acs4, Facl4 |
-342.5kb | Anti-sense | Chr. X - 113596247 - 113660024 on NCBI Build Rnor_6.0 |
| Downstream |
Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1 |
Ammecr1 | RGD1561004 | 127.2kb | Anti-sense | Chr. X - 114129834 - 114233011 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir652
Genomic Map