Assay Details
| Gene Symbol: Mir293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032760.1 | 1 | 39 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-294 | Mir294 | rno-mir-294 | -291.0 | Sense | Chr. 1 - 64536746 - 64536816 on NCBI Build Rnor_6.0 |
| Downstream | microRNA mir-291b | Mir291b | rno-mir-291b | 653.0 | Sense | Chr. 1 - 64537831 - 64537938 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir293
Genomic Map