Assay Details
| Gene Symbol: Mir291b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037337.1 | 1 | 59 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-293 | Mir293 | rno-mir-293 | -653.0 | Sense | Chr. 1 - 64537107 - 64537178 on NCBI Build Rnor_6.0 |
| Downstream | microRNA mir-292 | Mir292 | rno-mir-292 | 181.0 | Sense | Chr. 1 - 64538119 - 64538200 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir291b
Genomic Map