Assay Details
| Gene Symbol: Mir3583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037377.1 | 1 | 44 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DnaJ (Hsp40) homolog, subfamily C, member 8 |
Dnajc8 | Sense | Chr. 5 - 150725674 - 150743732 on NCBI Build Rnor_6.0 | ||
| Downstream | platelet-activating factor receptor | Ptafr | 13.6kb | Sense | Chr. 5 - 150746294 - 150775675 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3583
Genomic Map