Assay Details
| Gene Symbol: Mir3589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037387.1 | 1 | 61 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-505 | Mir505 | rno-mir-505 | Anti-sense | Chr. X - 143517014 - 143517080 on NCBI Build Rnor_6.0 | |
| Downstream | hypothetical protein LOC688842 | LOC688842 | 37.2kb | Anti-sense | Chr. X - 143554334 - 143596247 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3589
Genomic Map