Assay Details
| Gene Symbol: Mir26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031831.1 | 1 | 42 | 70 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CA339547.1 | - | 124 | Chr. - - [-] on Build | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 11 (proton-coupled divalent metal ion transporter), member 1 |
Slc11a1 |
Bcg, Itg, Lsh, Nramp1 |
-8.6kb | Sense | Chr. 9 - 81655610 - 81666697 on NCBI Build Rnor_6.0 |
| Downstream | villin 1 | Vil1 | Vil | 14.4kb | Sense | Chr. 9 - 81689802 - 81717623 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir26b
Genomic Map