Assay Details
| Gene Symbol: Nrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106036.2 | NP_001099506.1 | 1 - 2 | 381 | 64 | |
| XM_006251935.2 | XP_006251997.1 | 3 - 4 | 660 | 64 | ||
| XM_006251937.2 | XP_006251999.1 | 3 - 4 | 640 | 64 | ||
| XM_008770672.1 | XP_008768894.1 | 2 - 3 | 563 | 64 | ||
More Information
Set Membership: |
|

Nrl
Genomic Map