Assay Details
| Gene Symbol: Sall1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107415.2 | NP_001100885.2 | 1 - 2 | 102 | 70 | |
| XM_006255259.2 | XP_006255321.1 | 2 - 3 | 636 | 70 | ||
| XM_006255260.2 | XP_006255322.1 | 2 - 3 | 227 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sall1
Genomic Map