Assay Details
| Gene Symbol: Nek7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108346.2 | NP_001101816.1 | 6 - 7 | 595 | 63 | |
| XM_006249926.2 | XP_006249988.1 | 7 - 8 | 818 | 63 | ||
| XM_006249927.2 | XP_006249989.1 | 7 - 8 | 4436 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nek7
Genomic Map