Assay Details
| Gene Symbol: Lrit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191938.1 | NP_001178867.1 | 2 - 3 | 893 | 64 | |
| XM_006252787.2 | XP_006252849.1 | 2 - 3 | 2176 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrit2
Genomic Map