Assay Details
| Gene Symbol: Mir3559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037341.1 | 1 | 54 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uracil phosphoribosyltransferase (FUR1) homolog (S. cerevisiae) | Uprt | RGD1564806 | -50.7kb | Anti-sense | Chr. X - 75382567 - 75408539 on NCBI Build Rnor_6.0 |
| Downstream |
melanoma antigen, family E, 2 |
Magee2 | RGD1564872 | 278.4kb | Anti-sense | Chr. X - 75737724 - 75795202 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3559
Genomic Map