Assay Details
| Gene Symbol: Etv3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100992.2 | NP_001094462.2 | 2 - 3 | 312 | 63 | |
| XM_006232728.2 | XP_006232790.1 | 2 - 3 | 575 | 63 | ||
| XM_006232729.2 | XP_006232791.1 | 2 - 3 | 592 | 63 | ||
| XM_006232730.2 | XP_006232792.1 | 3 - 4 | 243 | 63 | ||
| XM_006232731.2 | XP_006232793.1 | 3 - 4 | 216 | 63 | ||
| GenBank mRNA | BC127525.1 | - | 2 - 3 | 312 | 7114610 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Etv3l
Genomic Map