Assay Details
| Gene Symbol: Dusp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108598.2 | NP_001102068.2 | 3 - 4 | 344 | 93 | |
| NM_001244784.1 | NP_001231713.1 | 3 - 4 | 344 | 93 | ||
| GenBank mRNA | BC168211.1 | - | 3 - 4 | 173 | 7135378 | 93 |
| CB576376.1 | - | 3 - 4 | 221 | 93 | ||
| CB585518.1 | - | 3 - 4 | 344 | 93 | ||
More Information
Set Membership: |
|

Dusp15
Genomic Map