Assay Details
| Gene Symbol: Hrh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073042.1 | 1 - 2 | 590 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosome-associated GTPase 2 | Mtg2 | Gtpbp5 | -13.5kb | Anti-sense | Chr. 3 - 175448313 - 175464109 on NCBI Build Rnor_6.0 |
| Downstream | oxysterol binding protein-like 2 | Osbpl2 | 14.3kb | Anti-sense | Chr. 3 - 175493664 - 175538965 on NCBI Build Rnor_6.0 |
More Information
Updated Mapping Information
Set Membership: |
|

Hrh3
Genomic Map