Assay Details
| Gene Symbol: LOC500148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073161.1 | 1 - 2 | 390 | 93 | ||
| GenBank mRNA | BC079164.1 | - | 1 - 2 | 390 | 7129532 | 93 |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | CK474108.1 | - | 389 | Chr. - - [-] on Build | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HECT and RLD domain containing E3 ubiquitin protein ligase family member 6 | Herc6 | -25.7kb | Sense | Chr. 4 - 88606851 - 88649421 on NCBI Build Rnor_6.0 | |
| Downstream |
protein phosphatase, Mg2+/Mn2+ dependent, 1K |
Ppm1k | RGD1308501 | 10.0kb | Anti-sense | Chr. 4 - 88694370 - 88721374 on NCBI Build Rnor_6.0 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC500148
Genomic Map