Assay Details
| Gene Symbol: Skp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007608.2 | NP_001007609.1 | 3 - 4 | 256 | 101 | |
| XM_006246244.2 | XP_006246306.1 | 3 - 4 | 555 | 101 | ||
| GenBank mRNA | BC058152.1 | - | 3 - 4 | 259 | 6890712 | 101 |
| FQ210698.1 | - | 2 - 3 | 154 | 101 | ||
| FQ212502.1 | - | 2 - 3 | 154 | 101 | ||
| FQ212528.1 | - | 2 - 3 | 154 | 101 | ||
| FQ212745.1 | - | 2 - 3 | 154 | 101 | ||
| FQ212916.1 | - | 2 - 3 | 154 | 101 | ||
| FQ213991.1 | - | 2 - 3 | 154 | 101 | ||
| FQ214002.1 | - | 2 - 3 | 154 | 101 | ||
| FQ214005.1 | - | 2 - 3 | 154 | 101 | ||
| FQ214369.1 | - | 2 - 3 | 154 | 101 | ||
| FQ214470.1 | - | 2 - 3 | 154 | 101 | ||
| FQ214982.1 | - | 2 - 3 | 154 | 101 | ||
| FQ215334.1 | - | 2 - 3 | 154 | 101 | ||
| FQ215452.1 | - | 2 - 3 | 154 | 101 | ||
| FQ219088.1 | - | 2 - 3 | 154 | 101 | ||
| FQ219401.1 | - | 2 - 3 | 154 | 101 | ||
| FQ224935.1 | - | 2 - 3 | 154 | 101 | ||
| FQ229076.1 | - | 2 - 3 | 154 | 101 | ||
| FQ230985.1 | - | 2 - 3 | 154 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Skp1
Genomic Map