Assay Details
| Gene Symbol: Zfp639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080907.2 | NP_001074376.1 | 4 - 5 | 458 | 95 | |
| XM_006232254.2 | XP_006232316.1 | 5 - 6 | 528 | 95 | ||
| XM_008760942.1 | XP_008759164.1 | 4 - 5 | 2007 | 95 | ||
| XM_008760943.1 | XP_008759165.1 | 5 - 6 | 1279 | 95 | ||
| XM_008760944.1 | XP_008759166.1 | 2 - 3 | 1049 | 95 | ||
| GenBank mRNA | BC087704.1 | - | 4 - 5 | 459 | 7313723 | 95 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_008760944.1 transcripts.
Set Membership: |
|

Zfp639
Genomic Map