Assay Details
Gene Symbol: Zfp639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080907.2 | NP_001074376.1 | 5 - 6 | 591 | 76 | |
XM_006232254.2 | XP_006232316.1 | 6 - 7 | 661 | 76 | ||
XM_008760942.1 | XP_008759164.1 | 5 - 6 | 2140 | 76 | ||
XM_008760943.1 | XP_008759165.1 | 6 - 7 | 1412 | 76 | ||
XM_008760944.1 | XP_008759166.1 | 3 - 4 | 1182 | 76 | ||
GenBank mRNA | BC087704.1 | - | 5 - 6 | 592 | 7313723 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
