Assay Details
| Gene Symbol: Rhbdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107067.2 | NP_001100537.2 | 3 - 4 | 377 | 67 | |
| NM_001270834.1 | NP_001257763.1 | 3 - 4 | 305 | 67 | ||
| GenBank mRNA | FQ230904.1 | - | 3 - 4 | 377 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhbdf2
Genomic Map