Assay Details
| Gene Symbol: Obp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128139.2 | NP_001121611.1 | 4 - 5 | 403 | 75 | |
| XM_008761608.1 | XP_008759830.1 | 4 - 5 | 1726 | 75 | ||
| GenBank mRNA | DQ537493.1 | - | 4 - 5 | 405 | 75 | |
More Information
Set Membership: |
|

Obp2a
Genomic Map