Assay Details
| Gene Symbol: Ppp1cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270983.1 | NP_001257912.1 | 3 - 4 | 666 | 83 | |
| NM_022498.2 | NP_071943.1 | 3 - 4 | 666 | 83 | ||
| XM_008769212.1 | XP_008767434.1 | 4 - 5 | 553 | 83 | ||
| GenBank mRNA | BC078825.1 | - | 3 - 4 | 593 | 7112668 | 83 |
| D90165.1 | - | 3 - 4 | 495 | 83 | ||
| D90166.1 | - | 3 - 4 | 609 | 83 | ||
| FN803249.1 | - | 3 - 4 | 553 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppp1cc
Genomic Map