Assay Details
| Gene Symbol: Cnnm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271050.1 | NP_001257979.1 | 3 - 4 | 1676 | 71 | |
| XM_006244827.2 | XP_006244889.1 | 3 - 4 | 1622 | 71 | ||
More Information
Set Membership: |
|

Cnnm4
Genomic Map