Assay Details
| Gene Symbol: Marcks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271090.2 | NP_001258019.1 | 1 - 2 | 484 | 61 | |
| GenBank mRNA | M59859.1 | - | 1 - 2 | 135 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Marcks
Genomic Map