Assay Details
| Gene Symbol: Ldlrad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271365.1 | NP_001258294.1 | 3 - 4 | 491 | 73 | |
| XM_008772172.1 | XP_008770394.1 | 2 - 3 | 361 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ldlrad4
Genomic Map