Assay Details
| Gene Symbol: Fabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012556.2 | NP_036688.1 | 3 - 4 | 375 | 78 | |
| GenBank mRNA | BC086947.1 | - | 3 - 4 | 375 | 7325407 | 78 |
| FQ209859.1 | - | 3 - 4 | 377 | 78 | ||
| FQ209983.1 | - | 3 - 4 | 384 | 78 | ||
| FQ210522.1 | - | 3 - 4 | 384 | 78 | ||
| FQ211310.1 | - | 3 - 4 | 384 | 78 | ||
| M10951.1 | - | 2 - 3 | 238 | 78 | ||
| M35991.1 | - | 3 - 4 | 376 | 78 | ||
| V01235.1 | - | 3 - 4 | 376 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fabp1
Genomic Map