Assay Details
| Gene Symbol: Ctsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013156.2 | NP_037288.1 | 2 - 3 | 197 | 63 | |
| GenBank mRNA | AB630323.1 | - | 1 - 2 | 138 | 63 | |
| BC063175.1 | - | 2 - 3 | 212 | 6918389 | 63 | |
| FQ209613.1 | - | 2 - 3 | 208 | 63 | ||
| FQ218649.1 | - | 2 - 3 | 206 | 63 | ||
| FQ228725.1 | - | 2 - 3 | 208 | 63 | ||
| FQ228844.1 | - | 2 - 3 | 208 | 63 | ||
| Y00697.1 | - | 2 - 3 | 201 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctsl
Genomic Map