Assay Details
| Gene Symbol: Ctsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013156.2 | NP_037288.1 | 6 - 7 | 846 | 83 | |
| GenBank mRNA | AB630323.1 | - | 5 - 6 | 787 | 83 | |
| BC063175.1 | - | 6 - 7 | 861 | 6918389 | 83 | |
| FQ209613.1 | - | 6 - 7 | 858 | 83 | ||
| FQ218649.1 | - | 6 - 7 | 855 | 83 | ||
| FQ228725.1 | - | 6 - 7 | 857 | 83 | ||
| Y00697.1 | - | 6 - 7 | 850 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctsl
Genomic Map