Assay Details
| Gene Symbol: Zfp354a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052798.2 | NP_434685.1 | 3 - 4 | 560 | 86 | |
| XM_006246227.2 | XP_006246289.1 | 4 - 5 | 1707 | 86 | ||
| XM_008767660.1 | XP_008765882.1 | 4 - 5 | 1704 | 86 | ||
| GenBank mRNA | BC078777.1 | - | 4 - 5 | 394 | 7127868 | 86 |
| M96548.1 | - | 3 - 4 | 562 | 86 | ||
More Information
Set Membership: |
|

Zfp354a
Genomic Map