Assay Details
| Gene Symbol: Spam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053967.2 | NP_446419.1 | 2 - 3 | 1277 | 84 | |
| GenBank mRNA | BC081748.1 | - | 2 - 3 | 1218 | 7134081 | 84 |
| X89999.1 | - | 2 - 3 | 1281 | 84 | ||
More Information
Set Membership: |
|

Spam1
Genomic Map