Assay Details
| Gene Symbol: Nacc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134413.2 | NP_599240.1 | 2 - 3 | 1038 | 96 | |
| XM_006255214.2 | XP_006255276.1 | 2 - 3 | 1032 | 96 | ||
| XM_008772346.1 | XP_008770568.1 | 2 - 3 | 1055 | 96 | ||
| GenBank mRNA | AF015911.1 | - | 3 - 4 | 1039 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nacc1
Genomic Map