Assay Details
| Gene Symbol: Dnase2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138539.2 | NP_612548.1 | 1 - 2 | 226 | 73 | |
| GenBank mRNA | AF178975.1 | - | 1 - 2 | 227 | 73 | |
| BC091124.1 | - | 1 - 2 | 129 | 7375398 | 73 | |
| FQ216710.1 | - | 1 - 2 | 226 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnase2
Genomic Map