Assay Details
| Gene Symbol: Dnase2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138539.2 | NP_612548.1 | 4 - 5 | 650 | 81 | |
| GenBank mRNA | AF178975.1 | - | 4 - 5 | 651 | 81 | |
| BC091124.1 | - | 4 - 5 | 553 | 7375398 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnase2
Genomic Map