Assay Details
| Gene Symbol: LOC100909675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110709.1 | 1 - 2 | 321 | 85 | ||
| GenBank mRNA | FN803941.1 | - | 1 - 2 | 321 | 85 | |
| FQ216719.1 | - | 1 - 2 | 53 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 126, member A |
Fam126a |
Drctnnb1a, RGD1562906 |
-98.5kb | Anti-sense | Chr. 4 - 7661717 - 7722750 on NCBI Build Rnor_6.0 |
| Downstream | translocase of outer mitochondrial membrane 7 homolog (yeast) | Tomm7 | 13.1kb | Anti-sense | Chr. 4 - 7835949 - 7842790 on NCBI Build Rnor_6.0 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100909675
Genomic Map