Assay Details
| Gene Symbol: Miat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_111959.1 | 4 - 5 | 1363 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
crystallin, beta A4 |
Cryba4 | -39.9kb | Sense | Chr. 12 - 50407843 - 50414432 on NCBI Build Rnor_6.0 | |
| Downstream | meningioma 1 | Mn1 | RGD1565571 | 752.3kb | Anti-sense | Chr. 12 - 51214192 - 51250230 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

Miat
Genomic Map